siRNA Details
Gene Symbol: Klk7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106254.1 | NP_001099724.1 | 2 | 270 | ||
XM_006228988.2 | XP_006229050.1 | 5 | 556 |

More Information
Other Species Targeted

Gene Ontology Categories:
