siRNA Details
Gene Symbol: LOC100911689 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748848.3 | XP_003748896.1 | 738 |
Gene Symbol: Klk1c10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135173.1 | NP_001128645.1 | 718 | |||
GenBank mRNA | S48142.1 | - | 4 | 675 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
