siRNA Details
Gene Symbol: Klk1c9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175759.2 | NP_786935.1 | 4 | 589 | ||
GenBank mRNA | BC061771.1 | - | 4 | 581 | 5621994 | |
M11566.1 | - | 4 | 589 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
