siRNA Details
Gene Symbol: Josd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106256.2 | NP_001099726.2 | 4 | 546 | ||
XM_006228996.2 | XP_006229058.1 | 5 | 828 | |||
XM_006228997.2 | XP_006229059.1 | 5 | 825 | |||
XM_006228998.2 | XP_006229060.1 | 4 | 767 | |||
GenBank mRNA | DV214517.1 | - | 4 | 546 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
