siRNA Details
Gene Symbol: Nosip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106260.1 | NP_001099730.1 | 4 | 293 | ||
XM_006229025.1 | XP_006229087.1 | 4 | 446 | |||
XM_006229026.2 | XP_006229088.1 | 5 | 478 | |||
GenBank mRNA | FQ216351.1 | - | 4 | 413 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
