siRNA Details
Gene Symbol: Hrc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181369.2 | NP_852034.1 | 4, 5 | 2274 | ||
GenBank mRNA | AY277279.1 | - | 4, 5 | 2274 | ||
FQ216289.1 | - | 4, 5 | 2368 | |||
FQ224148.1 | - | 4, 5 | 2360 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_010473, BC141121, AI322629, AF132218, AF158597.
Gene Ontology Categories:
