siRNA Details
Gene Symbol: Rasip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106261.2 | NP_001099731.1 | 3 | 669 | ||
XM_006229030.2 | XP_006229092.1 | 5 | 1754 | |||
GenBank mRNA | BC168876.1 | - | 3 | 669 | 7135603 |

More Information
Other Species Targeted

Gene Ontology Categories:
