siRNA Details
Gene Symbol: Syngr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025644.1 | NP_001020815.1 | 3 | 338 | ||
XM_006229034.2 | XP_006229096.1 | 3 | 554 | |||
XM_008759339.1 | XP_008757561.1 | 3 | 557 | |||
GenBank mRNA | BC098938.1 | - | 3 | 338 | 7455020 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
