siRNA Details
Gene Symbol: Otog | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106262.1 | NP_001099732.1 | 30 | 3591 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_013624, U96411.
Gene Ontology Categories:
