siRNA Details
Gene Symbol: Dbx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009644.1 | NP_001009644.1 | 1 | 121 | ||
GenBank mRNA | AB197138.1 | - | 1 | 121 |

More Information
Other Species Targeted

Gene Ontology Categories:
