siRNA Details
Gene Symbol: Olig3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106269.1 | NP_001099739.1 | 1 | 338 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_053008, AB038698, BC057564.
Gene Ontology Categories:
