siRNA Details
Gene Symbol: Map7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106270.2 | NP_001099740.2 | 11 | 1576 | ||
NM_001198638.1 | NP_001185567.1 | 9 | 1109 | |||
GenBank mRNA | CK480167.1 | - | 2 | 297 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
