siRNA Details
Gene Symbol: Cpeb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106276.1 | NP_001099746.1 | 7 | 1092 | ||
XM_008759509.1 | XP_008757731.1 | 7 | 1701 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_007755, NM_001252525, Y08260, AK136088, BC125476.
Gene Ontology Categories:
