siRNA Details
Gene Symbol: Fam154b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106277.1 | NP_001099747.1 | 3 | 660 | ||
XM_006229473.2 | XP_006229535.1 | 4 | 871 | |||
XM_006229474.2 | XP_006229536.1 | 4 | 2563 | |||
XM_006229475.2 | XP_006229537.1 | 5 | 954 | |||
XM_008759511.1 | XP_008757733.1 | 5 | 1762 |
