siRNA Details
Gene Symbol: Olr13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000116.1 | NP_001000116.1 | 1 | 790 |
Gene Symbol: LOC100909966 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748916.2 | XP_003748964.1 | 802 |
