siRNA Details
Gene Symbol: Eed | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106278.2 | NP_001099748.1 | 8 | 1237 | ||
XM_006229664.2 | XP_006229726.1 | 8 | 1016 | |||
GenBank mRNA | BC168872.1 | - | 8 | 1237 | 9025004 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
