siRNA Details
Gene Symbol: Crebzf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271791.1 | NP_001258720.1 | 1 | 1312 | ||
XM_006229688.2 | XP_006229750.1 | 1 | 1677 | |||
XM_008759646.1 | XP_008757868.1 | 1 | 1677 | |||
GenBank mRNA | FN797334.1 | - | 1 | 361 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_145151, AF408398, BG094109, CX237465.
Gene Ontology Categories:
