siRNA Details
Gene Symbol: Tmem126a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011557.1 | NP_001011557.1 | 3 | 247 | ||
XM_006229689.2 | XP_006229751.1 | 3 | 412 | |||
GenBank mRNA | BC089104.1 | - | 3 | 247 | 7307750 | |
FQ210414.1 | - | 3 | 268 | |||
FQ220582.1 | - | 3 | 406 | |||
FQ231598.1 | - | 3 | 297 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
