siRNA Details
Gene Symbol: Tmem126a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011557.1 | NP_001011557.1 | 2, 3 | 171 | ||
XM_006229689.2 | XP_006229751.1 | 2, 3 | 336 | |||
GenBank mRNA | BC089104.1 | - | 2, 3 | 171 | 7307750 | |
FQ210414.1 | - | 2, 3 | 192 | |||
FQ220582.1 | - | 2, 3 | 330 | |||
FQ231598.1 | - | 2, 3 | 221 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
