siRNA Details
Gene Symbol: Olr35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000121.1 | NP_001000121.1 | 1 | 851 | ||
GenBank mRNA | BC086526.1 | - | 865 | 7303669 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
