siRNA Details
Gene Symbol: RGD1311703 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006230086.2 | XP_006230148.1 | 5 | 549 | ||
GenBank mRNA | BC089924.1 | - | 5 | 514 | 7307309 | |
FM053223.1 | - | 5 | 553 | |||
FQ225875.1 | - | 5 | 549 |
