siRNA Details
Gene Symbol: Olr196 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000187.1 | NP_001000187.1 | 1 | 592 |
Gene Symbol: Olr194 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000948.1 | NP_001000948.1 | 1 | 592 |

More Information
Other Species Targeted

Gene Ontology Categories:
