siRNA Details
Gene Symbol: Olr206 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000193.1 | NP_001000193.1 | 1 | 588 |
Gene Symbol: Olr204 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000192.1 | NP_001000192.1 | 1 | 588 |
