siRNA Details
Gene Symbol: Olr233 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000207.1 | NP_001000207.1 | 1 | 487 |
Gene Symbol: Olr232 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001035.1 | NP_001001035.1 | 1 | 487 |
