siRNA Details
Gene Symbol: Olr239 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000211.1 | NP_001000211.1 | 1 | 229 |
Gene Symbol: Olr235 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000209.1 | NP_001000209.1 | 1 | 229 |

More Information
Other Species Targeted

Gene Ontology Categories:
