siRNA Details
Gene Symbol: Olr267 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000228.1 | NP_001000228.1 | 1 | 627 |
Gene Symbol: Olr264 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000999.1 | NP_001000999.1 | 1 | 627 |
