siRNA Details
Gene Symbol: Vwa3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079885.1 | NP_001073354.1 | 3 | 259 | ||
NM_001198653.1 | NP_001185582.1 | 3 | 259 | |||
XM_006230159.2 | XP_006230221.1 | 3 | 1663 | |||
GenBank mRNA | BC128762.1 | - | 3 | 259 | 7113062 |
