siRNA Details
Gene Symbol: Fam57b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106296.2 | NP_001099766.2 | 2 | 351 | ||
XM_006230237.2 | XP_006230299.1 | 3 | 427 | |||
XM_006230239.2 | XP_006230301.1 | 2 | 172 | |||
XM_006230240.2 | XP_006230302.1 | 2 | 947 | |||
GenBank mRNA | BC161920.1 | - | 2 | 351 | 8370175 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
