siRNA Details
Gene Symbol: Pagr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013901.1 | NP_001013923.1 | 1 | 760 | ||
GenBank mRNA | BC088203.1 | - | 1 | 760 | 7379842 |

More Information
Other Species Targeted

Gene Ontology Categories:
