siRNA Details
Gene Symbol: Prkacb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077645.1 | NP_001071113.1 | 10 | 1065 | ||
XM_006233451.2 | XP_006233513.1 | 10 | 1782 | |||
XM_006233452.2 | XP_006233514.1 | 10 | 3287 | |||
XM_006233453.2 | XP_006233515.1 | 10 | 1529 | |||
GenBank mRNA | D10770.1 | - | 10 | 1065 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
