siRNA Details
Gene Symbol: LOC102554302 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006230372.1 | XP_006230434.1 | 3 | 732 | ||
XM_006230373.1 | XP_006230435.1 | 2 | 618 |

More Information
Other Species Targeted

