siRNA Details
Gene Symbol: Ahsp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106299.1 | NP_001099769.1 | 3 | 274 | ||
GenBank mRNA | BC157822.1 | - | 3 | 198 | 8374205 | |
FQ225822.1 | - | 3 | 195 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
