siRNA Details
Gene Symbol: Bag3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011936.1 | NP_001011936.1 | 4 | 1477 | ||
GenBank mRNA | BC085857.1 | - | 4 | 1477 | 7190249 | |
DQ631552.1 | - | 4 | 1480 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_013863, AB041583, AK171734, AK016510, AF130471, and others.
Gene Ontology Categories:
