siRNA Details
Gene Symbol: Fam175b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106307.1 | NP_001099777.2 | 9 | 965 | ||
GenBank mRNA | FQ218234.1 | - | 9 | 954 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
