siRNA Details
Gene Symbol: Fuom | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106310.1 | NP_001099780.1 | 6 | 851 | ||
GenBank mRNA | FQ212771.1 | - | 4 | 772 |
Gene Symbol: LOC100911225 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748887.3 | XP_003748935.1 | 6 | 473 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
