siRNA Details
Gene Symbol: Olr300 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000237.1 | NP_001000237.1 | 1 | 763 | ||
XM_008759976.1 | XP_008758198.1 | 3 | 2743 | |||
XM_008759977.1 | XP_008758199.1 | 3 | 2870 | |||
XM_008759978.1 | XP_008758200.1 | 3 | 2832 |
