siRNA Details
Gene Symbol: Olr307 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000241.1 | NP_001000241.1 | 1 | 823 |
Gene Symbol: LOC103690271 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760020.1 | XP_008758242.1 | 1 | 823 |
