siRNA Details
Gene Symbol: Sirt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106313.2 | NP_001099783.1 | 3 | 367 | ||
XM_006230526.2 | XP_006230588.1 | 3 | 447 | |||
XM_006230529.2 | XP_006230591.1 | 3 | 1061 | |||
XM_008759979.1 | XP_008758201.1 | 3 | 487 | |||
XM_008759980.1 | XP_008758202.1 | 3 | 487 | |||
XM_008759981.1 | XP_008758203.1 | 3 | 355 | |||
GenBank mRNA | BC167006.1 | - | 3 | 367 | 7314283 | |
EU886468.1 | - | 3 | 393 | |||
FQ213357.1 | - | 2 | 193 | |||
FQ218084.1 | - | 3 | 398 |
