siRNA Details
Gene Symbol: Irf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033691.1 | NP_001028863.1 | 9 | 1727 | ||
XM_006230540.1 | XP_006230602.1 | 10 | 1826 | |||
XM_006230541.1 | XP_006230603.1 | 10 | 1730 | |||
GenBank mRNA | BC104686.1 | - | 9 | 1727 | 7442863 | |
FQ232130.1 | - | 8 | 1182 | |||
FQ232859.1 | - | 7 | 1019 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
