siRNA Details
Gene Symbol: Ppfia1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106320.2 | NP_001099790.2 | 22 | 3082 | ||
XM_006230715.2 | XP_006230777.1 | 26 | 3278 | |||
XM_006230717.2 | XP_006230779.1 | 25 | 3248 | |||
XM_006230718.2 | XP_006230780.1 | 26 | 3278 | |||
XM_006230719.2 | XP_006230781.1 | 25 | 3215 | |||
XM_006230720.2 | XP_006230782.1 | 25 | 3203 | |||
XM_006230721.2 | XP_006230783.1 | 24 | 3188 | |||
XM_006230722.2 | XP_006230784.1 | 22 | 3083 | |||
XM_006230723.2 | XP_006230785.1 | 22 | 3083 | |||
XM_006230724.2 | XP_006230786.1 | 24 | 3140 |
Gene Symbol: LOC103689951 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749028.3 | XP_003749076.2 | 5 | 584 | ||
XM_006230626.2 | XP_006230688.1 | 4 | 557 | |||
XM_006230627.2 | XP_006230689.1 | 5 | 585 | |||
XM_006230628.2 | XP_006230690.1 | 4 | 520 | |||
XM_006230629.2 | XP_006230691.1 | 3 | 491 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
