siRNA Details
Gene Symbol: Ppfia1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106320.2 | NP_001099790.2 | 15 | 2022 | ||
XM_006230715.2 | XP_006230777.1 | 16 | 2098 | |||
XM_006230717.2 | XP_006230779.1 | 16 | 2098 | |||
XM_006230718.2 | XP_006230780.1 | 16 | 2098 | |||
XM_006230719.2 | XP_006230781.1 | 16 | 2098 | |||
XM_006230720.2 | XP_006230782.1 | 15 | 2023 | |||
XM_006230721.2 | XP_006230783.1 | 16 | 2098 | |||
XM_006230722.2 | XP_006230784.1 | 15 | 2023 | |||
XM_006230723.2 | XP_006230785.1 | 15 | 2023 | |||
XM_006230724.2 | XP_006230786.1 | 15 | 2023 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
