siRNA Details
Gene Symbol: VDAC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135694.2 | NP_001129166.1 | 8 | 598 | ||
NM_005662.6 | NP_005653.3 | 7 | 595 | |||
XM_006716394.1 | XP_006716457.1 | 6 | 682 | |||
GenBank mRNA | AF038962.1 | - | 7 | 484 | ||
AK058201.1 | - | 7 | 469 | |||
AK307224.1 | - | 6 | 342 | |||
BC056870.1 | - | 7 | 480 | 6163890 | ||
U90943.1 | - | 5 | 385 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
