siRNA Details
Gene Symbol: Lrp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106321.2 | NP_001099791.2 | 15 | 3482 | ||
XM_006230726.2 | XP_006230788.1 | 15 | 3538 | |||
GenBank mRNA | BC166786.1 | - | 15 | 3482 | 7935737 |
