siRNA Details
Gene Symbol: Tcirg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199089.2 | NP_954520.2 | 14, 15 | 1894 | ||
XM_006230727.2 | XP_006230789.1 | 15, 16 | 1998 | |||
XM_006230728.2 | XP_006230790.1 | 15, 16 | 1969 | |||
XM_008760097.1 | XP_008758319.1 | 15, 16 | 2065 | |||
GenBank mRNA | DQ286426.1 | - | 14, 15 | 1894 |
