siRNA Details
Gene Symbol: Ndufv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006972.1 | NP_001006973.1 | 7 | 1075 | ||
GenBank mRNA | BC083709.1 | - | 7 | 1075 | 7190583 | |
FQ214688.1 | - | 7 | 1083 | |||
FQ217076.1 | - | 7 | 1133 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
