siRNA Details
Gene Symbol: Rbm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170484.1 | NP_001163955.1 | 3 | 665 | ||
XM_008760101.1 | XP_008758323.1 | 3 | 1589 | |||
GenBank mRNA | FQ212306.1 | - | 3 | 771 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_009032, U89506, BC130256, AK049659, BC144949, and others.
Gene Ontology Categories:
