siRNA Details
Gene Symbol: Brms1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009605.1 | NP_001009605.1 | 396 | |||
GenBank mRNA | BC088469.1 | - | 396 | 7128111 |
Gene Symbol: LOC100911841 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749038.3 | XP_003749086.1 | 4 | 456 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
