siRNA Details
Gene Symbol: Sf3b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106326.1 | NP_001099796.2 | 3, 4 | 319 | ||
XM_008760105.1 | XP_008758327.1 | 3, 4 | 310 | |||
GenBank mRNA | FQ220434.1 | - | 3, 4 | 290 |
