siRNA Details
Gene Symbol: Efemp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005907.2 | NP_001005907.1 | 10 | 1248 | ||
NM_001277341.1 | NP_001264270.1 | 11 | 1371 | |||
NM_001277346.1 | NP_001264275.1 | 9 | 1041 | |||
XM_006230742.2 | XP_006230804.1 | 11 | 1711 | |||
XM_006230743.1 | XP_006230805.1 | 11 | 1362 | |||
XM_006230744.2 | XP_006230806.1 | 11 | 1368 | |||
GenBank mRNA | BC083804.1 | - | 10 | 1186 | 7109182 |
