siRNA Details
Gene Symbol: Cdc42bpg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130013.1 | NP_001123485.1 | 35 | 4732 | ||
XM_006230751.2 | XP_006230813.1 | 35 | 4671 | |||
XM_006230752.2 | XP_006230814.1 | 35 | 4598 | |||
XM_008760108.1 | XP_008758330.1 | 35 | 4382 |
